siRNA Details
Gene Symbol: Wbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029066.1 | NP_083342.1 | 3 | 274 | ||
GenBank mRNA | AK015863.1 | - | 3 | 305 | ||
BC119520.2 | - | 3 | 274 | 40055925 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029066.1 | NP_083342.1 | 3 | 274 | ||
GenBank mRNA | AK015863.1 | - | 3 | 305 | ||
BC119520.2 | - | 3 | 274 | 40055925 |