siRNA Details
| Gene Symbol: Wbp2nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029066.1 | NP_083342.1 | 2, 3 | 192 | ||
| GenBank mRNA | AK015863.1 | - | 2, 3 | 223 | ||
| BC119520.2 | - | 2, 3 | 192 | 40055925 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wbp2nl| Gene Symbol: Wbp2nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029066.1 | NP_083342.1 | 2, 3 | 192 | ||
| GenBank mRNA | AK015863.1 | - | 2, 3 | 223 | ||
| BC119520.2 | - | 2, 3 | 192 | 40055925 | ||