siRNA Details
| Gene Symbol: Tmem114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029070.2 | NP_083346.1 | 2 | 725 | ||
| GenBank mRNA | AK015757.1 | - | 2 | 726 | ||
| BC027071.1 | - | 2 | 698 | |||
| BC147190.1 | - | 2 | 502 | 9056234 | ||
| BC147191.1 | - | 2 | 502 | 9056214 | ||

Tmem114
Genomic Map