siRNA Details
| Gene Symbol: Trim14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029077.4 | NP_083353.1 | 2, 3 | 359 | ||
| GenBank mRNA | AB117644.1 | - | 2, 3 | 370 | ||
| AK017887.1 | - | 2, 3 | 333 | |||
| AK076277.1 | - | 341 | ||||
| AK133220.1 | - | 2, 3 | 282 | |||
| AK148837.1 | - | 2, 3 | 323 | |||
| AK172159.1 | - | 2, 3 | 142 | |||
| AK172895.1 | - | 2, 3 | 338 | |||
| BC048627.1 | - | 110 | ||||
| BC116738.1 | - | 2, 3 | 358 | 40126057 | ||
| BC119130.1 | - | 2, 3 | 358 | 8733879 | ||

Trim14
Genomic Map