siRNA Details
| Gene Symbol: Pcf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029078.3 | NP_083354.3 | 2, 3 | 618 | ||
| GenBank mRNA | AK161260.1 | - | 2, 3 | 642 | ||
| AK161614.1 | - | 2, 3 | 608 | |||
| AK173035.1 | - | 2, 3 | 590 | |||
| BC150781.1 | - | 2, 3 | 454 | 9087692 | ||

Pcf11
Genomic Map