siRNA Details
| Gene Symbol: Ddit4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029083.2 | NP_083359.1 | 3 | 1116 | ||
| GenBank mRNA | AK017926.1 | - | 3 | 1116 | ||
| BC132645.1 | - | 3 | 1115 | 40130922 | ||
| BC145370.1 | - | 3 | 1115 | 9053928 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddit4| Gene Symbol: Ddit4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029083.2 | NP_083359.1 | 3 | 1116 | ||
| GenBank mRNA | AK017926.1 | - | 3 | 1116 | ||
| BC132645.1 | - | 3 | 1115 | 40130922 | ||
| BC145370.1 | - | 3 | 1115 | 9053928 | ||