siRNA Details
| Gene Symbol: Yipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205157.1 | NP_001192086.1 | 4 | 351 | ||
| NM_001205158.1 | NP_001192087.1 | 4 | 341 | |||
| NM_138303.2 | NP_612176.1 | 3 | 489 | |||
| GenBank mRNA | AA967152.1 | - | 3 | 288 | ||
| AK004959.1 | - | 3 | 275 | |||
| AK033705.1 | - | 4 | 300 | |||
| AK133631.1 | - | 4 | 311 | |||
| BC002282.1 | - | 3 | 404 | 3497525 | ||
| BC003289.1 | - | 4 | 282 | 3583087 | ||

Yipf2
Genomic Map