siRNA Details
| Gene Symbol: Hhatl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029095.2 | NP_083371.2 | 10 | 1378 | ||
| GenBank mRNA | AK003605.1 | - | 10 | 1255 | ||
| AK157924.1 | - | 10 | 1176 | |||
| BC024464.1 | - | 10 | 1234 | 4975643 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hhatl| Gene Symbol: Hhatl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029095.2 | NP_083371.2 | 10 | 1378 | ||
| GenBank mRNA | AK003605.1 | - | 10 | 1255 | ||
| AK157924.1 | - | 10 | 1176 | |||
| BC024464.1 | - | 10 | 1234 | 4975643 | ||