siRNA Details
| Gene Symbol: Wipi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178398.4 | NP_848485.1 | 4 | 528 | ||
| GenBank mRNA | AK010881.1 | - | 4 | 556 | ||
| AK043857.1 | - | 4 | 561 | |||
| AK155113.1 | - | 4 | 558 | |||
| AK169051.1 | - | 4 | 502 | |||
| AK172030.1 | - | 4 | 504 | |||
| BC044894.1 | - | 4 | 553 | 4235237 | ||

Wipi2
Genomic Map