siRNA Details
| Gene Symbol: Glt8d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029102.3 | NP_083378.2 | 3 | 254 | ||
| GenBank mRNA | AK003894.1 | - | 3 | 255 | ||
| AK040842.1 | - | 3 | 222 | |||
| AK086276.1 | - | 2 | 1923 | |||
| AK136351.1 | - | 3 | 235 | |||
| BC082561.1 | - | 3 | 183 | 30603848 | ||

Glt8d2
Genomic Map