siRNA Details
| Gene Symbol: Usp38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027554.2 | NP_081830.2 | 9 | 2571 | ||
| GenBank mRNA | AK014740.1 | - | 3 | 675 | ||
| AK028904.1 | - | 9 | 2550 | |||
| AK049287.1 | - | 9 | 2539 | |||
| AK052219.1 | - | 9 | 2594 | |||
| AK054116.1 | - | 9 | 2573 | |||
| AK142200.1 | - | 9 | 2566 | |||
| BC054404.1 | - | 9 | 2571 | 6494448 | ||
| BC057122.1 | - | 9 | 2561 | 6834543 | ||
| BC058784.1 | - | 9 | 2499 | 6310341 | ||

Usp38
Genomic Map