siRNA Details
| Gene Symbol: Kbtbd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029116.2 | NP_083392.1 | 2 | 2748 | ||
| GenBank mRNA | AK015509.1 | - | 1 | 295 | ||
| AK046634.1 | - | 1 | 1820 | |||
| AK050286.1 | - | 1 | 1604 | |||
| AK080001.1 | - | 1 | 1767 | |||
| AK129197.1 | - | 1 | 1711 | |||
| BC080858.1 | - | 1 | 1719 | 6852896 | ||

Kbtbd11
Genomic Map