siRNA Details
| Gene Symbol: 4930480E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177966.2 | NP_001171437.1 | 1 | 346 | ||
| GenBank mRNA | AK015598.1 | - | 1 | 346 | ||
| BC116859.1 | - | 1 | 346 | 40126178 | ||
| BC116861.1 | - | 1 | 346 | 40126180 | ||

4930480E11Rik
Genomic Map