siRNA Details
| Gene Symbol: Iqca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029122.2 | NP_083398.2 | 4 | 695 | ||
| GenBank mRNA | AK015512.1 | - | 4 | 696 | ||
| AK016353.1 | - | 4 | 696 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqca| Gene Symbol: Iqca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029122.2 | NP_083398.2 | 4 | 695 | ||
| GenBank mRNA | AK015512.1 | - | 4 | 696 | ||
| AK016353.1 | - | 4 | 696 | |||