siRNA Details
| Gene Symbol: Iqca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029122.2 | NP_083398.2 | 13, 14 | 1784 | ||
| GenBank mRNA | AK139726.1 | - | 6, 7 | 639 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqca| Gene Symbol: Iqca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029122.2 | NP_083398.2 | 13, 14 | 1784 | ||
| GenBank mRNA | AK139726.1 | - | 6, 7 | 639 | ||