siRNA Details
| Gene Symbol: 1700049E17Rik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006737.1 | - | 2 | 259 | ||
| Gene Symbol: 4930503E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029131.3 | NP_083407.1 | 2 | 301 | ||
| GenBank mRNA | AK015686.1 | - | 2 | 301 | ||

1700049E17Rik1
Genomic Map