siRNA Details
| Gene Symbol: Usp47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133758.3 | NP_598519.2 | 23, 24 | 3668 | ||
| NM_177249.3 | NP_796223.2 | 24, 25 | 3728 | |||
| GenBank mRNA | AK030909.1 | - | 8, 9 | 1619 | ||
| AK131138.1 | - | 14, 15 | 2345 | |||
| BC024117.1 | - | 7, 8 | 1494 | |||
| BC057362.1 | - | 4, 5 | 752 | |||
| BC089364.1 | - | 4, 5 | 542 | |||

Usp47
Genomic Map