siRNA Details
| Gene Symbol: Rab11fip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033172.3 | NP_001028344.2 | 1, 2 | 728 | ||
| NM_001164367.1 | NP_001157839.1 | 1, 2 | 156 | |||
| GenBank mRNA | AK015534.1 | - | 1, 2 | 157 | ||
| AK154916.1 | - | 1, 2 | 728 | |||
| BC139380.1 | - | 1, 2 | 491 | 8862402 | ||
| BC139419.1 | - | 1, 2 | 491 | 8862441 | ||

Rab11fip2
Genomic Map