siRNA Details
| Gene Symbol: Sf3a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029157.3 | NP_083433.1 | 17 | 1493 | ||
| GenBank mRNA | AK030743.1 | - | 17 | 1528 | ||
| AK150733.1 | - | 17 | 1491 | |||
| AK151141.1 | - | 17 | 1491 | |||
| AK151382.1 | - | 17 | 1491 | |||
| AK161646.1 | - | 17 | 1514 | |||
| AK168966.1 | - | 17 | 1517 | |||
| AK169324.1 | - | 17 | 1537 | |||
| AK186857.1 | - | 3 | 180 | |||
| AK210925.1 | - | 3 | 138 | |||
| AK213278.1 | - | 3 | 163 | |||
| AK216219.1 | - | 3 | 180 | |||
| BC009141.1 | - | 17 | 1493 | 3598647 | ||
| BC092058.1 | - | 17 | 1525 | 4017388 | ||

Sf3a3
Genomic Map