siRNA Details
| Gene Symbol: Uhrf1bp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029166.2 | NP_083442.2 | 6 | 816 | ||
| GenBank mRNA | AB093261.1 | - | 6 | 781 | ||
| AK033300.1 | - | 7 | 824 | |||
| AK087280.1 | - | 4 | 368 | |||
| BB529788.2 | - | 2 | 208 | |||
| BC030383.1 | - | 775 | ||||
| BC132134.1 | - | 6 | 609 | 40130411 | ||
| BC138092.1 | - | 6 | 609 | 8861111 | ||

Uhrf1bp1l
Genomic Map