siRNA Details
| Gene Symbol: Ube2dnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081661.1 | NP_001075130.1 | 2 | 648 | ||
| GenBank mRNA | AK015883.1 | - | 2 | 648 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2dnl2| Gene Symbol: Ube2dnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081661.1 | NP_001075130.1 | 2 | 648 | ||
| GenBank mRNA | AK015883.1 | - | 2 | 648 | ||