siRNA Details
| Gene Symbol: Rprd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081293.1 | NP_001074762.1 | 2 | 492 | ||
| GenBank mRNA | AK012860.1 | - | 2 | 367 | ||
| AK015972.1 | - | 2 | 403 | |||
| AK083674.1 | - | 2 | 315 | |||
| AK087157.1 | - | 2 | 318 | |||
| AK129150.1 | - | 2 | 171 | |||
| BC067054.1 | - | 2 | 297 | 30542928 | ||

Rprd2
Genomic Map