siRNA Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029186.2 | NP_083462.2 | 8 | 1321 | ||
| GenBank mRNA | AK015988.1 | - | 5 | 677 | ||
| AK031554.1 | - | 8 | 1267 | |||
| AK052797.1 | - | 4 | 307 | |||
| AK151797.1 | - | 8 | 1296 | |||
| AK161305.1 | - | 8 | 1277 | |||
| AK170247.1 | - | 1268 | ||||
| BC026385.1 | - | 5 | 1277 | |||
| BC031169.1 | - | 7 | 1094 | |||
| BC049142.1 | - | 5 | 607 | |||
| BC058746.1 | - | 8 | 1267 | 6399588 | ||

Tmem180
Genomic Map