siRNA Details
| Gene Symbol: Ankrd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167757.1 | NP_001161229.1 | 3 | 554 | ||
| NM_029202.2 | NP_083478.2 | 3 | 551 | |||
| GenBank mRNA | AK015948.1 | - | 3 | 495 | ||
| AK077080.1 | - | 3 | 523 | |||
| BC049731.1 | - | 3 | 551 | 6771078 | ||

Ankrd7
Genomic Map