siRNA Details
| Gene Symbol: Sv2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029210.1 | NP_083486.1 | 11 | 1981 | ||
| GenBank mRNA | AK015921.1 | - | 4 | 486 | ||
| AK137053.1 | - | 7 | 823 | |||
| AK173092.1 | - | 11 | 1981 | |||
| BC132111.1 | - | 10 | 1817 | 40130388 | ||
| BC137861.1 | - | 10 | 1817 | 8860880 | ||

Sv2c
Genomic Map