siRNA Details
| Gene Symbol: Prr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282012.1 | NP_001268941.1 | 4 | 696 | ||
| NM_001282013.1 | NP_001268942.1 | 6 | 701 | |||
| NM_001282024.1 | NP_001268953.1 | 5 | 795 | |||
| NM_145487.3 | NP_663462.2 | 4 | 724 | |||
| GenBank mRNA | AJ504719.1 | - | 4 | 1493 | ||
| AK132945.1 | - | 4 | 424 | |||
| BU148736.1 | - | 6 | 658 | |||

Prr3
Genomic Map