siRNA Details
Gene Symbol: Rnf19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029219.1 | NP_083495.1 | 9 | 1747 | ||
GenBank mRNA | AK015966.1 | - | 4 | 431 | ||
AK183889.1 | - | 3 | 272 | |||
BC052529.1 | - | 3 | 349 | |||
BC117807.1 | - | 9 | 1305 | |||
BC139808.1 | - | 9 | 1308 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
