siRNA Details
Gene Symbol: 4930564B18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029230.1 | NP_083506.1 | 4 | 613 | ||
GenBank mRNA | AK016216.1 | - | 4 | 614 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930564B18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029230.1 | NP_083506.1 | 4 | 613 | ||
GenBank mRNA | AK016216.1 | - | 4 | 614 |