siRNA Details
| Gene Symbol: Slc35f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029238.2 | NP_083514.1 | 3 | 747 | ||
| GenBank mRNA | AK034308.1 | - | 3 | 749 | ||
| AK043617.1 | - | 3 | 765 | |||
| AK046093.1 | - | 3 | 957 | |||
| AK047949.1 | - | 3 | 744 | |||
| BC138981.1 | - | 3 | 646 | 8862003 | ||
| BC145720.1 | - | 3 | 646 | 40131023 | ||

Slc35f4
Genomic Map