siRNA Details
| Gene Symbol: Prkd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171004.1 | NP_001164475.1 | 15 | 2660 | ||
| NM_001171005.1 | NP_001164476.1 | 14 | 1856 | |||
| NM_029239.3 | NP_083515.2 | 15 | 2657 | |||
| GenBank mRNA | AK030058.1 | - | 14 | 1853 | ||
| BC027320.1 | - | 14 | 2059 | |||
| BC037012.1 | - | 14 | 2301 | 5342428 | ||
| BC089164.1 | - | 15 | 2559 | 5400250 | ||

Prkd3
Genomic Map