siRNA Details
| Gene Symbol: Prkd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171004.1 | NP_001164475.1 | 11 | 2240 | ||
| NM_001171005.1 | NP_001164476.1 | 10 | 1436 | |||
| NM_029239.3 | NP_083515.2 | 11 | 2240 | |||
| GenBank mRNA | AK030058.1 | - | 10 | 1433 | ||
| BC027320.1 | - | 10 | 1639 | |||
| BC037012.1 | - | 10 | 1884 | 5342428 | ||
| BC089164.1 | - | 11 | 2139 | 5400250 | ||

Prkd3
Genomic Map