siRNA Details
| Gene Symbol: Ankrd53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029245.3 | NP_083521.2 | 3 | 605 | ||
| GenBank mRNA | AK016222.1 | - | 2 | 401 | ||
| AK133099.1 | - | 3 | 607 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd53| Gene Symbol: Ankrd53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029245.3 | NP_083521.2 | 3 | 605 | ||
| GenBank mRNA | AK016222.1 | - | 2 | 401 | ||
| AK133099.1 | - | 3 | 607 | |||