siRNA Details
| Gene Symbol: 4930550C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029247.3 | NP_083523.1 | 6 | 534 | ||
| GenBank mRNA | AK016083.1 | - | 6 | 535 | ||
| AK153618.1 | - | 4 | 431 | |||
| BC115638.1 | - | 4 | 435 | 40102982 | ||
| BC115639.1 | - | 6 | 522 | 40102983 | ||

4930550C14Rik
Genomic Map