siRNA Details
| Gene Symbol: Taf1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027261.3 | NP_081537.1 | 3 | 255 | ||
| NM_029248.2 | NP_083524.2 | 3 | 255 | |||
| GenBank mRNA | AK012656.1 | - | 1 | 105 | ||
| AK012662.1 | - | 3 | 207 | |||
| AK014920.1 | - | 3 | 207 | |||
| AK016109.1 | - | 3 | 209 | |||
| AK020036.1 | - | 2 | 167 | |||
| AK050165.1 | - | 3 | 211 | |||
| AK161656.1 | - | 3 | 210 | |||
| AK166329.1 | - | 3 | 210 | |||
| AK166633.1 | - | 3 | 210 | |||
| BC027813.1 | - | 1 | 43 | |||
| BC029906.1 | - | 2 | 710 | |||
| BC056964.1 | - | 2 | 193 | |||
| BC110660.1 | - | 2 | 197 | 3158156 | ||
| BC117068.1 | - | 2 | 187 | 40126387 | ||
| BC117070.1 | - | 2 | 187 | 40126389 | ||
| BU850945.1 | - | 3 | 259 | |||

Taf1d
Genomic Map