siRNA Details
| Gene Symbol: Etnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029250.2 | NP_083526.2 | 7 | 1230 | ||
| GenBank mRNA | AK016135.1 | - | 7 | 1212 | ||
| AK033268.1 | - | 7 | 1207 | |||
| AK044502.1 | - | 7 | 1231 | |||
| AK049827.1 | - | 7 | 944 | |||

Etnk1
Genomic Map