siRNA Details
| Gene Symbol: 4930550L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023774.3 | NP_076263.1 | 2 | 828 | ||
| GenBank mRNA | AF319984.1 | - | 1 | 678 | ||
| AK016090.1 | - | 2 | 802 | |||
| BC087920.1 | - | 2 | 828 | 30281980 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930550L24Rik| Gene Symbol: 4930550L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023774.3 | NP_076263.1 | 2 | 828 | ||
| GenBank mRNA | AF319984.1 | - | 1 | 678 | ||
| AK016090.1 | - | 2 | 802 | |||
| BC087920.1 | - | 2 | 828 | 30281980 | ||