siRNA Details
| Gene Symbol: Spp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029269.2 | NP_083545.1 | 2 | 286 | ||
| GenBank mRNA | AJ315513.1 | - | 2 | 247 | ||
| AK002814.1 | - | 2 | 287 | |||
| BC027494.1 | - | 2 | 212 | 1433655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spp2| Gene Symbol: Spp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029269.2 | NP_083545.1 | 2 | 286 | ||
| GenBank mRNA | AJ315513.1 | - | 2 | 247 | ||
| AK002814.1 | - | 2 | 287 | |||
| BC027494.1 | - | 2 | 212 | 1433655 | ||