siRNA Details
| Gene Symbol: NDUFA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005002.4 | NP_004993.1 | 7 | 697 | ||
| GenBank mRNA | AF050641.1 | - | 7 | 687 | ||
| AK291067.1 | - | 7 | 674 | |||
| AK308588.1 | - | 8 | 796 | |||
| BC003351.1 | - | 6 | 551 | |||
| BC009311.2 | - | 7 | 678 | 4106931 | ||
| BC015837.1 | - | 7 | 693 | 4293690 | ||
| BC111546.1 | - | 7 | 669 | 5761015 | ||
| L04490.1 | - | 7 | 663 | |||

NDUFA9
Genomic Map