siRNA Details
| Gene Symbol: Secisbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012732.1 | - | 6, 7 | 939 | ||
| AK030021.1 | - | 6, 7 | 909 | |||
| AK037988.1 | - | 6, 7 | 939 | |||
| AK156076.1 | - | 6, 7 | 985 | |||
| AK168634.1 | - | 6, 7 | 942 | |||
| BC125391.1 | - | 6, 7 | 965 | 40129906 | ||
| BC125393.1 | - | 6, 7 | 962 | 40129908 | ||
| BC144795.1 | - | 6, 7 | 960 | |||

Secisbp2
Genomic Map