siRNA Details
| Gene Symbol: Secisbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012732.1 | - | 5 | 682 | ||
| AK030021.1 | - | 5 | 652 | |||
| AK037988.1 | - | 5 | 682 | |||
| AK156076.1 | - | 5 | 728 | |||
| AK168634.1 | - | 5 | 685 | |||
| BC125391.1 | - | 5 | 708 | 40129906 | ||
| BC125393.1 | - | 5 | 705 | 40129908 | ||
| BC144795.1 | - | 5 | 703 | |||

Secisbp2
Genomic Map