siRNA Details
| Gene Symbol: Mettl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029280.4 | NP_083556.2 | 1 | 331 | ||
| GenBank mRNA | AA466649.1 | - | 1 | 329 | ||
| AK013063.1 | - | 1 | 314 | |||
| AK133594.1 | - | 1 | 321 | |||
| BC027547.1 | - | 1 | 264 | 1531973 | ||
| CJ063020.1 | - | 1 | 321 | |||

Mettl5
Genomic Map