siRNA Details
| Gene Symbol: NDUFA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005002.4 | NP_004993.1 | 9, 10 | 926 | ||
| GenBank mRNA | AF050641.1 | - | 9, 10 | 916 | ||
| AK291067.1 | - | 9, 10 | 903 | |||
| BC003351.1 | - | 8, 9 | 780 | |||
| BC009311.2 | - | 9, 10 | 907 | 4106931 | ||
| BC015837.1 | - | 9, 10 | 922 | 4293690 | ||
| BC111546.1 | - | 9, 10 | 898 | 5761015 | ||
| L04490.1 | - | 9, 10 | 892 | |||

NDUFA9
Genomic Map