siRNA Details
| Gene Symbol: Igfbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018741.2 | NP_061211.1 | 3 | 633 | ||
| GenBank mRNA | AB006141.1 | - | 3 | 621 | ||
| AK012705.1 | - | 3 | 635 | |||
| AK013339.1 | - | 2 | 159 | |||
| BC051960.1 | - | 3 | 603 | 5710646 | ||

Igfbpl1
Genomic Map