siRNA Details
| Gene Symbol: Fam183b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162878.1 | NP_001156350.1 | 2 | 234 | ||
| NM_029283.1 | NP_083559.1 | 3 | 294 | |||
| GenBank mRNA | AK013920.1 | - | 2 | 237 | ||
| AK157972.1 | - | 3 | 273 | |||
| BC048748.1 | - | 3 | 251 | |||
| BC061254.1 | - | 3 | 244 | |||
| BC094630.1 | - | 3 | 251 | |||
| BC117741.2 | - | 3 | 350 | |||
| BC126964.1 | - | 3 | 347 | |||
| FJ468008.1 | - | 3 | 233 | |||

Fam183b
Genomic Map