siRNA Details
| Gene Symbol: 1700001G01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005589.1 | - | 518 | |||
| AK133007.1 | - | 4 | 620 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001G01Rik| Gene Symbol: 1700001G01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005589.1 | - | 518 | |||
| AK133007.1 | - | 4 | 620 | |||