siRNA Details
| Gene Symbol: 1700006E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029287.1 | NP_083563.1 | 4 | 403 | ||
| GenBank mRNA | AK005669.1 | - | 4 | 407 | ||
| BC100517.1 | - | 4 | 378 | |||
| BC116819.1 | - | 4 | 399 | 40126138 | ||
| BC116821.1 | - | 4 | 399 | 40126140 | ||

1700006E09Rik
Genomic Map