siRNA Details
Gene Symbol: Syce3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162880.1 | NP_001156352.1 | 2 | 179 | ||
NM_001162882.1 | NP_001156354.1 | 2 | 147 | |||
GenBank mRNA | AK005702.1 | - | 2 | 101 | ||
BC048615.1 | - | 2 | 179 | |||
EU079025.1 | - | 2 | 140 | |||
HQ130280.1 | - | 1 | 74 |
