siRNA Details
| Gene Symbol: Dynlrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029297.1 | NP_083573.1 | 2 | 99 | ||
| GenBank mRNA | AK005789.1 | - | 2 | 99 | ||
| BC048623.1 | - | 2 | 140 | 6743790 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dynlrb2| Gene Symbol: Dynlrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029297.1 | NP_083573.1 | 2 | 99 | ||
| GenBank mRNA | AK005789.1 | - | 2 | 99 | ||
| BC048623.1 | - | 2 | 140 | 6743790 | ||