siRNA Details
Gene Symbol: Iqcf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029300.1 | NP_083576.1 | 2 | 433 | ||
GenBank mRNA | AK005734.1 | - | 2 | 434 | ||
BC125546.1 | - | 2 | 392 | 40130061 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Iqcf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029300.1 | NP_083576.1 | 2 | 433 | ||
GenBank mRNA | AK005734.1 | - | 2 | 434 | ||
BC125546.1 | - | 2 | 392 | 40130061 |