siRNA Details
| Gene Symbol: 1700009N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081095.1 | NP_001074564.1 | 2 | 565 | ||
| GenBank mRNA | BC050973.1 | - | 2 | 341 | ||
| BC116915.1 | - | 2 | 414 | 40126234 | ||
| BC116917.1 | - | 2 | 414 | 40126236 | ||

1700009N14Rik
Genomic Map